Learn More
Wheaton™ Vaccum Filter with Frit
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Brand:  Wheaton™ 419452
Code : A0
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11751883
  • £690.00 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Specifications
Specifications
Large Sample Volumes, Liquids With A Heavy Particulate Load And Viscous Liquids When A High Flow Rate Is Required
Vaccum Filter
90 mm