missing translation for 'onlineSavingsMsg'
Learn More

Vaccinia Virus A33R Antibody - (Copenhagen), Novus Biologicals™

Rabbit Polyclonal Antibody

£154.00 - £302.00

Specifications

Antigen Vaccinia Virus A33R
Dilution Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30281681
View Documents Promotion Details
Novus Biologicals
NBP3-43775-20ul
20 μL
£154.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30281680
View Documents Promotion Details
Novus Biologicals
NBP3-43775-100ul
100 μL
£302.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vaccinia Virus A33R Polyclonal antibody specifically detects Vaccinia Virus A33R in Virus samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Vaccinia Virus A33R
ELISA, Western Blot
Unconjugated
Rabbit
Virus
OPG161
IgG
Antigen and protein A Affinity-purified
Western Blot 1:2000-1:20000, ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 μm filtered solution in PBS
Produced in rabbits immunized with purified, recombinant Vaccinia Virus A33R (Copenhagen) (Uniprot#: P68616; Val57-Asn185)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.