missing translation for 'onlineSavingsMsg'
Learn More

VABP Antibody (6C239.4), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18423987
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423987 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423987 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224876AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VABP Monoclonal antibody specifically detects VABP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VABP
Applications Western Blot
Classification Monoclonal
Clone 6C239.4
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein
Host Species Mouse
Immunogen Full length recombinant protein was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30820
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.