missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VABP Antibody (6C239.4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24876AF594
This item is not returnable.
View return policy
Description
VABP Monoclonal antibody specifically detects VABP in Human samples. It is validated for Western Blot
Specifications
| VABP | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 6C239.4 | |
| Western Blot | |
| A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein | |
| Full length recombinant protein was used as the immunogen. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 30820 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction