Learn More
Invitrogen™ V5 Tag Monoclonal Antibody, Alexa Fluor™ 647

Description
The Alexa FluorTM dyes provide for extraordinarily bright antibody conjugates. At the time of preparation, the products are certified to be free of unconjugated dyes and are tested in a cytological experiment to ensure low nonspecific staining. For Alexa FluorTM 647 the approximate fluorescence excitation (Ex) and emission (Em) maxima, in nm: Ex 650/Em 668.
Specifications
Specifications
| Antigen | V5 Tag |
| Applications | Inhibition Assays |
| Classification | Monoclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | PBS with 2mM sodium azide; pH 7.5 |
| Gene Alias | simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag |
| Host Species | Mouse |
| Immunogen | sequence -Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-. |
| Purification Method | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.