missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ V5 Tag Monoclonal Antibody (SV5-Pk1)

Product Code. 10347642
Change view
Click to view available options
Quantity:
50 μL
500 μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10347642 50 μL Each
10605353 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10347642 Supplier Invitrogen™ Supplier No. R96025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

NOTE: R960-25 was previously sold under product #46-0705. When re-ordering, please use #R960-25. R960CUS was previously sold under product #46-1157. When re-ordering, please use #R960CUS. This antibody recognizes amino acid sequence: -Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-. This antibody is functionally tested against 20 ng of an E. coli expressed fusion protein containing a V5 epitope using a chemiluminescent substrate at a 1 minute exposure. This antibody has also been tested in Western blot against 25 ng of recombinant Positope™ protein. The Positope™ control protein is a 53 kDa recombinant protein that contains seven epitope tags, including His (C-term), HisG, c-myc, and V5. Low background was observed using chemiluminescent or alkaline phosphatase reagents for detection. For Western blot, dilute in PBS or Tris-Buffered Saline (TBS) containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM or TBSTM). Using chemiluminescence as the detection method, no cross-reactivity has been observed in bacterial lysates. In mammalian lysates, a few cross-reactive proteins have been observed upon overexposure of blots. This antibody has also been used successfully to immunoprecipitate fusion proteins that contain theV5 epitope.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen V5 Tag
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SV5-Pk1
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide
Gene Alias simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
Host Species Mouse
Immunogen V5 synthetic peptide: Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-.
Purification Method Purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.