missing translation for 'onlineSavingsMsg'
Learn More

V26 Standard: Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V,W, Zn @ 500 μg/g in 75 cSt Hydrocarbon Oil

Product Code. 30318149
Change view
Click to view available options
Quantity:
400 g
Unit Size:
400g
Product Code. Quantity unitSize
30318149 400 g 400g
1 options
This item is not returnable. View return policy

Product Code. 30318149

Brand: VHG Labs VHGV26+CO+W500400G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Antimony∣Barium∣Bismuth∣Boron∣Cadmium∣Calcium∣Chromium∣Cobalt∣Copper∣Indium∣Iron∣Lead∣Lithium∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silicon∣Silver∣Sodium∣Tin∣Titanium∣Tungsten∣Vanadium∣Zinc
CAS 7429-90-5, 7440-36-0, 7440-39-3, 7440-69-9, 7440-42-8, 7440-43-9
Analyte or Component Names Aluminium∣Antimony∣Barium∣Bismuth∣Boron∣Cadmium∣Calcium∣Chromium∣Cobalt∣Copper∣Indium∣Iron∣Lead∣Lithium∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silicon∣Silver∣Sodium∣Tin∣Titanium∣Tungsten∣Vanadium∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix 75 cSt Hydrocarbon Oil
Industry Type Industrial QC
Quantity 400 g
Concentration or Composition (by Analyte or Components) Aluminium 500 μg/g∣Antimony 500 μg/g∣Barium 500 μg/g∣Bismuth 500 μg/g∣Boron 500 μg/g∣Cadmium 500 μg/g∣Calcium 500 μg/g∣Chromium 500 μg/g∣Cobalt 500 μg/g∣Copper 500 μg/g∣Indium 500 μg/g∣Iron 500 μg/g∣Lead 500 μg/g∣Lithium 500 μg/g∣Magnesium 500 μg/g∣Manganese 500 μg/g∣Molybdenum 500 μg/g∣Nickel 500 μg/g∣Phosphorus 500 μg/g∣Potassium 500 μg/g∣Silicon 500 μg/g∣Silver 500 μg/g∣Sodium 500 μg/g∣Tin 500 μg/g∣Titanium 500 μg/g∣Tungsten 500 μg/g∣Vanadium 500 μg/g∣Zinc 500 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.