missing translation for 'onlineSavingsMsg'
Learn More

V23 Standard: Al, Ba, B, Cd, Ca, Co, Cr, Cu, Fe, K, Li, Pb, Mg, Mn, Mo, Ni, P, Sb, Si, Ag, Na, Sn, Ti, V, Zn @ 100 μg/g in 75 cSt Hydrocarbon Oil

Product Code. 30321840
Change view
Click to view available options
Quantity:
400 g
Unit Size:
400g
Product Code. Quantity unitSize
30321840 400 g 400g
1 options
This item is not returnable. View return policy

Product Code. 30321840

Brand: VHG Labs VHGV23+LI+CO100400G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium∣Antimony∣Barium∣Boron∣Cadmium∣Calcium∣Chromium∣Cobalt∣Copper∣Iron∣Lead∣Lithium∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silicon∣Silver∣Sodium∣Tin∣Titanium∣Vanadium∣Zinc
CAS 7429-90-5, 7440-36-0, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2
Analyte or Component Names Aluminium∣Antimony∣Barium∣Boron∣Cadmium∣Calcium∣Chromium∣Cobalt∣Copper∣Iron∣Lead∣Lithium∣Magnesium∣Manganese∣Molybdenum∣Nickel∣Phosphorus∣Potassium∣Silicon∣Silver∣Sodium∣Tin∣Titanium∣Vanadium∣Zinc
Shipping Condition Room Temperature
Solution Type Mixture
Element Multi-element
Solvent or Matrix 75 cSt Hydrocarbon Oil
Industry Type Industrial QC
Quantity 400 g
Concentration or Composition (by Analyte or Components) Aluminium 100 μg/g∣Antimony 100 μg/g∣Barium 100 μg/g∣Boron 100 μg/g∣Cadmium 100 μg/g∣Calcium 100 μg/g∣Chromium 100 μg/g∣Cobalt 100 μg/g∣Copper 100 μg/g∣Iron 100 μg/g∣Lead 100 μg/g∣Lithium 100 μg/g∣Magnesium 100 μg/g∣Manganese 100 μg/g∣Molybdenum 100 μg/g∣Nickel 100 μg/g∣Phosphorus 100 μg/g∣Potassium 100 μg/g∣Silicon 100 μg/g∣Silver 100 μg/g∣Sodium 100 μg/g∣Tin 100 μg/g∣Titanium 100 μg/g∣Vanadium 100 μg/g∣Zinc 100 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.