missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V1a Vasopressin R/AVPR1A Antibody (612130), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10932CL1
This item is not returnable.
View return policy
Description
V1a Vasopressin R/AVPR1A Monoclonal antibody specifically detects V1a Vasopressin R/AVPR1A in Human samples. It is validated for Flow Cytometry
Specifications
| V1a Vasopressin R/AVPR1A | |
| Monoclonal | |
| CoraFluor 1 | |
| Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor | |
| NS0 mouse myeloma cell line transfected with human V1a Vasopressin R/AVPR1A, Accession # P37288 | |
| 0.1 mL | |
| GPCR | |
| 552 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 612130 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction