missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vectair™ V-Fresh™ Air Freshener Pad
Click to view available options
Color:
Green
Red
Description
- Freshens air and neutralises odours
- VOC exempt, environmentally aware
- No propellants or solvents
- Non-hazardous - no restrictions in transport and no storage issues
- Fixes to wide variety of hard, smooth surfaces
- Integrated hanging slot means it can also be hung from a variety of objects, including on the side of toilet bowls, urinals, tiled walls or glass windows
- Subtle yet effective
- Simple to install and replace
Specifications
Specifications
| Scent | Cucumber Melon |
| Autoclavable | Not Autoclavable |
| Color | Green |
| Description | Made of EVA (Ethylene Vinyl Acetate Copolymer) |
| For Use With (Application) | Solid state universal air freshener |
| Quantity | 12 Pk. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction