missing translation for 'onlineSavingsMsg'
Learn More

Vectair™ V-Fresh™ Air Freshener Pad

Product Code. 15802825
Encompass_Preferred
Click to view available options
Color:
Green
Red
This item is not returnable. View return policy

Product Code. 15802825

Brand: Vectair™ VFRESH RED

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Vectair™ V-Fresh™ Air Freshener Pad is a solid state universal air freshener with added malodour eliminating micro porous materials to combat bad odours.

  • Freshens air and neutralises odours
  • VOC exempt, environmentally aware
  • No propellants or solvents
  • Non-hazardous - no restrictions in transport and no storage issues
  • Fixes to wide variety of hard, smooth surfaces
  • Integrated hanging slot means it can also be hung from a variety of objects, including on the side of toilet bowls, urinals, tiled walls or glass windows
  • Subtle yet effective
  • Simple to install and replace

Specifications

Scent Apple Orchard
Autoclavable Not Autoclavable
Color Red
Description Made of EVA (Ethylene Vinyl Acetate Copolymer)
For Use With (Application) Solid state universal air freshener
Quantity 12 Pk.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.