missing translation for 'onlineSavingsMsg'
Learn More

TCR V beta 8.1/8.2 Monoclonal Antibody (KJ16-133 (KJ-16, KJI6, KJ-I6)), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

£79.00 - £105.00

Specifications

Antigen TCR V beta 8.1/8.2
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15258269
View Documents
Invitrogen
11-5813-81
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15268269
GREENER_CHOICE
View Documents
Invitrogen™
11-5813-82
100 μg
£105.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This KJ16-133 monoclonal antibody recognizes mouse T cell receptors (TCR) containing Vbeta 8.1 or 8.2, which is expressed by 15-20% of peripheral T cells in most strains of mice (ie. Balb/c, AKR, C57BL/6, B10.A) and by a number of T cell hybridomas (ie. DO-11.10, 3DT-52.5, D1G10G11, SKK-9.11, SKK-45.10, MDK16, S18.4). Applications Reported: This KJ16-133 (KJ-16, KJI6, KJ-I6) antibody has been reported for use in flow cytometric analysis. Applications Tested: This KJ16 (KJ-16, KJI6, KJ-I6) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease. Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
Specifications

Specifications

TCR V beta 8.1/8.2
Flow Cytometry
FITC
Rat
Mouse
Primary
4° C, store in dark, DO NOT FREEZE!
0.5 mg/mL
Monoclonal
Liquid
RUO
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.