missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ V-ATPase G3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575698
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ATP6V1G3 (V-ATPase G3) is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
Specifications
| V-ATPase G3 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1G3 | |
| ATP6G3; ATP6V1G3; ATPase H+ transporting V1 subunit G3; ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3; ATPase, H+ transporting, lysosomal V1 subunit G3; ATPase, H+ transporting, lysosomal, V1 subunit G3; ATPase, H+ transporting, V1 subunit G, isoform 3; vacuolar ATP synthase subunit G 3; vacuolar proton pump G subunit 3; vacuolar proton pump subunit G 3; vacuolar proton pump, subunit G3; V-ATPase 13 kDa subunit 3; V-ATPase G subunit 3; V-ATPase G3 subunit; V-ATPase subunit G 3; Vma10; V-type proton ATPase subunit G 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127124, 289407, 338375 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BMC1, Q96LB4 | |
| ATP6V1G3 | |
| Synthetic peptide corresponding to residues in Human V-ATPase G3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction