missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ V-ATPase G3 Monoclonal Antibody (3A5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523000
This item is not returnable.
View return policy
Description
Peptide Sequence: EEAMVEIDQY RMQRDKEFRL KQSKIMGSQN NLSDEIEEQT LGKIQELNGH YNKYMESVMN QLLSMVCDMK PEIHVNYRAT N.
ATP6V1G3 (V-ATPase G3) is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
Specifications
| V-ATPase G3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q96LB4 | |
| ATP6V1G3 | |
| ATP6V1G3 (NP_573569, 38 a.a. ~ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3A5 | |
| Unconjugated | |
| ATP6V1G3 | |
| ATP6G3; ATP6V1G3; ATPase H+ transporting V1 subunit G3; ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3; ATPase, H+ transporting, lysosomal V1 subunit G3; ATPase, H+ transporting, lysosomal, V1 subunit G3; ATPase, H+ transporting, V1 subunit G, isoform 3; vacuolar ATP synthase subunit G 3; vacuolar proton pump G subunit 3; vacuolar proton pump subunit G 3; vacuolar proton pump, subunit G3; V-ATPase 13 kDa subunit 3; V-ATPase G subunit 3; V-ATPase G3 subunit; V-ATPase subunit G 3; Vma10; V-type proton ATPase subunit G 3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 127124 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction