missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ V-ATPase G1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565342

Product Code. 15968094

  • £471.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AEIEQYRLQRE KEFKAKEAAA LGSRGSCSTE VEKETQEKMT ILQTYFRQNR DEVLDNLLAF VCDIRPEIHE NYR Highest antigen sequence identity to the following orthologs - mouse 92%, rat 95%.

This protein is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

V-ATPase G1
Polyclonal
Unconjugated
ATP6V1G1
1810024D14Rik; AA960677; ATP6G; ATP6G1; ATP6GL; ATP6J; ATP6V1G1; ATPase H+ transporting V1 subunit G1; ATPase, H transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J; ATPase, H+ transporting, lysosomal 13kD, V1 subunit G; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1; ATPase, H+ transporting, lysosomal V1 subunit G1; ATPase, H+ transporting, V1 subunit G; DKFZp547P234; lysosomal 13kDa; vacuolar ATP synthase subunit M16; vacuolar H(+)-ATPase subunit G 1; vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16; VAG1; V-ATPase 13 kDa subunit 1; V-ATPase subunit G 1; Vma10; V-type proton ATPase subunit G 1
Rabbit
Antigen affinity chromatography
RUO
9550
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75348
ATP6V1G1
Recombinant Human V-ATPase G1. Recombinant protein control fragment (Product #RP-104763).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.