missing translation for 'onlineSavingsMsg'
Learn More

V alpha 24 J alpha 18 TCR Antibody (6B11), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18427862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427862 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427862 Supplier Novus Biologicals Supplier No. NBP200267JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

V alpha 24 J alpha 18 TCR Monoclonal antibody specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Flow (Cell Surface), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen V alpha 24 J alpha 18 TCR
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), Flow Cytometry (Cell Surface)
Classification Monoclonal
Clone 6B11
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell
Host Species Mouse
Immunogen Peptide epitope in the CDR3 region of V alpha 24
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.