missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ V 9302 hydrochloride
Click to view available options
:
10 mg
Description
V 9302 hydrochloride is an alanine-serine-cysteine transporter 2 (ASCT2, encoded by gene SLC1A5) inhibitor. It inhibits glutamate uptake (IC50 = 9.6 μM in HEK-293 cells). It reduces cancer cell growth and proliferation, also increases cell death and oxidative stress in vitro and in vivo. V 9302 hydrochloride inhibits mTORC1 activity, mitochondrial respiration, proliferation and migration of vascular smooth muscle cells (VSMCs).
Specifications
Specifications
| Chemical Name or Material | (2S)-2-Amino-4-[bis[[2-[(3-methylphenyl)methoxy]phenyl]methyl]amino]butanoic acid hydrochloride |
| CAS | 2416138-42-4 |
| Quantity | 10 mg |
| Target | Other Ion Pump/Transporter Inhibitors |
| Molecular Formula | C34H38N2O4.HCl |
| Purity | 0.97 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction