missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vβ 11 T-Cell Receptor Rat anti-Mouse, BUV615, Clone: RR3-15, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 751297
This item is not returnable.
View return policy
Description
Vβ 11 T-Cell Receptor Monoclonal antibody specifically detects Vβ 11 T-Cell Receptor in Mouse samples. It is validated for Flow Cytometry
Specifications
| Vβ 11 T-Cell Receptor | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| TCR V beta 11 | |
| Mouse Cytolytic T-Cell Clone OH6 | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| RR3-15 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction