missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UXT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11047-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms.Specifications
| UXT | |
| Polyclonal | |
| Unconjugated | |
| UXT | |
| ART 27, Protein UXT, UXT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22294, 299313, 8409 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q63ZY7, Q9UBK9, Q9WTZ0 | |
| UXT | |
| UXT Fusion Protein Ag1513 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction