missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UXT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563566
This item is not returnable.
View return policy
Description
Immunogen sequence: LKFIDRKSSL LTELSNSLTK DSMNIKAHIH MLLEGLRELQ GLQNFPEKPH H Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
This gene encodes a novel protein which is highly conserved in mouse. It interacts with the N-terminus of the androgen receptor and plays a role in facilitating receptor-induced transcriptional activation. It is also likely to be involved in tumorigenesis as it is abundantly expressed in tumor tissues. This gene is part of a gene cluster on chromosome Xp11.23. Alternative splicing results in 2 transcript variants encoding different isoforms.
Specifications
| UXT | |
| Polyclonal | |
| Unconjugated | |
| UXT | |
| 0910002B17Rik; androgen receptor trapped clone 27 protein; ART-27; HSPC024; Protein UXT; RP1-212G6.2; SKP2-associated alpha PFD 1; STAP1; ubiquitously expressed prefoldin like chaperone; ubiquitously expressed transcript; ubiquitously expressed transcript protein; ubiquitously-expressed transcript; ubiquitously-expressed, prefoldin-like chaperone; Unknown (protein for MGC:133475); UXT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBK9 | |
| UXT | |
| Recombinant protein corresponding to Human UXT. Recombinant protein control fragment (Product #RP-101746). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction