Learn More
UVP White Light Replacement Tubes
Product Code. 12893918
Change view
Click to view available options
For Use With (Equipment):
14 Type EL series UV lamp
16 Type EL series UV Lamp, High Performance Transilluminator
Model MRL-58 Multiple-ray Lamp
Model TW-43 White Light Transilluminator
Models TLW-20, TMW-20, TW-26 Transilluminator
Wattage:
4 W
6 W
8 W
14 W
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Wattage unitSize
12893918 Model MRL-58 Multiple-ray Lamp 8 W Each
12802989 Model TW-43 White Light Transilluminator 14 W Each
12873918 Models TLW-20, TMW-20, TW-26 Transilluminator 8 W Each
12822979 14 Type EL series UV lamp 4 W Each
12882979 16 Type EL series UV Lamp, High Performance Transilluminator 6 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12893918 Supplier UVP Supplier No. 34005602
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Light Source White Daylight
Wattage 8 W
For Use With (Equipment) Model MRL-58 Multiple-ray Lamp
Includes 8 W warm white tube
Packaging Quantity 1 Ea.
Product Type Replacement Tube