missing translation for 'onlineSavingsMsg'
Learn More

UVP GFP Filters for Colony Doc-It Imaging Station

Product Code. 12852939
Change view
Click to view available options
Wavelength:
513 to 557nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
12852939 513 to 557nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12852939 Supplier UVP Supplier No. 38034001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Two position

Specifications

For Use With (Application) Used for colony counting
For Use With (Equipment) Uvp™ colony doc-it imaging station
Includes 50mm Sq Filter
Quantity Each
Size 50mm Square
Wavelength 513 to 557nm
Product Type Gfp Filter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.