Learn More
UVP Filter Assemblies for Handheld EL Series UV Lamps
Brand:  UVP 38-0158-04
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12872949
  • £133.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Specifications
White Light/Longwave Filter Assembly
Model UVL-18 handheld EL-series UV lamp
Filter Assembly
1 Ea.