Learn More
UVP Filter Assemblies for Handheld EL Series UV Lamps
Brand: UVP 38-0158-02
Code : 04
Additional Details : Weight : 0.10000kg
Specifications
Shortwave Filter Assembly | |
Model UVS-28 and UMV-28 handheld EL-series UV lamp | |
Filter Assembly | |
1 Ea. |