missing translation for 'onlineSavingsMsg'
Learn More

UVP Clear UV Transmitting Covers

Product Code. 12843848
Change view
Click to view available options
For Use With (Equipment):
xx-15 series benchtop uv lamps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12843848 xx-15 series benchtop uv lamps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12843848 Supplier UVP Supplier No. 19014001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Clear
For Use With (Equipment) xx-15 series benchtop uv lamps
Includes Plastic cover for UVP xx-15 series benchtop uv lamps
Quantity Each
Product Type UV Transmitting Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.