Learn More
UVP Camera Filter SYBR Green
Allows visualization of fluorophores from 460-470nm
Brand:  UVP 38-0219-01
Code : 04
Additional Details : Weight : 0.04888kg
Product Code. 12810703
  • £305.00 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Blocks UV and IR radiation emitted from the transilluminator
Specifications
Specifications
50
For use with chromadoc imaging system
Camera Filter Sybr Green