Learn More
UVP Lamp Funnel for Blak-Ray™ B-100AP High-Intensity UV Inspection Lamps
Product Code. 12813029
Change view
Click to view available options
For Use With (Equipment):
Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
12813029 Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp Each
1 options
Product Code. 12813029 Supplier UVP Supplier No. 19012601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for B-100AP High-Intensity UV Inspection Lamps
Lamp funnel for use with B-100AP, B-100AP/R, B-100SP high intensity UV inspection lamps.
Specifications
For Use With (Application) Directs UV source to specific area
For Use With (Equipment) Blak-Ray Model B-100AP, B-100AP/R, B-100SP, B-100YP High Intensity UV Lamp
Material Metal
Packaging Quantity 1 Ea.
Product Type Lamp Funnel