missing translation for 'onlineSavingsMsg'
Learn More

UVP BL-15 UV Inspection Lamp Replacement Tube

Longwave 365nm BLB UV lamp is ideal for inspection applications in hard to reach places.

Brand:  UVP 34-0017-01

Code : 04

Additional Details : Weight : 0.08000kg

 View more versions of this product

Product Code. 12853848

  • £40.45 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

15 W
UVP™ BL-15 UV inspection lamp
434.8 mm
365 nm
17.12 in.
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers