missing translation for 'onlineSavingsMsg'
Learn More

UVI 3003, MedChemExpress

Product Code. 16494138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494138

Brand: Medchem Express HY107500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively.

UVI 3003 is a highly selective antagonist of retinoid X receptor (RXR), and inhibits xenopus and human RXRα in Cos7 cells, with IC50s of 0.22 and 0.24 μM, respectively.

Specifications

Chemical Name or Material UVI 3003
CAS 847239-17-2
Color White
Purity Grade Notes Research
Molecular Formula C28H36O4
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (229.05 mM; Need ultrasonic)
SMILES CCCCCOC1=C(C2=CC(/C=C/C(O)=O)=CC=C2O)C=C3C(C(C)(CCC3(C)C)C)=C1
Molecular Weight (g/mol) 436.58
Formula Weight 436.58
Percent Purity 99.03%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.