Learn More
Honeywell™ Uvex™ Xenova Hygiene Slipper S2 SRC
Product Code. 15358062 Shop All Uvex Products
Click to view available options
Size:
36
43
Product Code. 15358062
Brand: Honeywell™ 6962.8/36
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Especially light metal-free shoe in slipper shape
  • Number of seams limited to the minimum necessary to make it difficult to set impurities
  • Optimized comfort thanks to uvex climazone technology
  • Breathable, high-tech microfibre upper, abrasion-resistant and hydrophobic
  • Breathable distance mesh lining material
  • uvex anti-twist rear cap for more stability and kink protection
  • Suitable for allergy sufferers as made of synthetic materials
  • uvex xenova™ duo toe caps in duo component technology, thermally insulating Soft padded shaft end and padded tongue
  • Interchangeable comfort footbed, antistatic, with moisture transport system
  • uvex 3D hydroflex™ foam footbed for optimal cushioning in the heel and forefoot area
  • Meets the ESD requirement: leakage resistance <35 megohms
  • PUR outsole
  • Standard EN ISO 20345: 2011 S2 SRC
  • Size 35 - 50
  • TRUSTED_SUSTAINABILITY
    Specifications
    Size 36
    Width (Metric) 11 mm