Learn More
Honeywell™ Uvex™ Xenova Hygiene Shoe S2
Product Code. 15368092 Shop All Uvex Products
Change view
Click to view available options
Size:
39
44
48
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
15368092 48
15368082 39
15328092 44
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15368092 Supplier Honeywell™ Supplier No. 6964.8/48
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Ultra light lace-up shoe
  • Uvex xenova™ hygiene Shoe 6964 S2 SRC
  • Ultra light lace-up shoe
  • Metal-free
  • Minimal number of seams to reduce staining
  • Uvex climazone technology for improved comfort
  • High-tech breathable microfibre material upper, abrasion-resistant, water-resistant
  • Ultra breathable distance mesh lining uvex anti-twist heel cap for greater stability and protection from ankle twisting
  • Made with synthetic textiles - suitable for people allergic to chrome
  • Uvex xenova™ duo toecap with duo component technology
  • thermally insulated
  • Soft padded collar and tongue
  • Ultra comfortable removable insole, antistatic, with a moisture control system
  • Uvex 3D hydroflex™ foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Fulfils ESD requirements: resistance to ground < 35 megaohms
  • PUR outsole
  • Standard EN ISO 20345:2011 S2 SRC
  • Colour white (Sizes 35 - 50)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance ESD certification, EN ISO 20345:2011 S2 SRC
    Color White, Gray
    Design Low, lace-up
    Size 48
    Toe Style Xenova™ duo toecap with duo component technology