Learn More
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves
Product Code. 17220421 Shop All Uvex Products
Change view
Click to view available options
Size:
6
8
9
10
11
Unit Size:
10 pairs
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17220421 6 10 pairs
15335662 8 Each
15345662 9 Each
15355662 10 Each
15365662 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17220421 Supplier uvex Supplier No. 6024806
  • ONLINE EXCLUSIVE PRICE
    £19.35
    £17.38 / 10 pairs
    Save £1.97
    10% Off
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Unipur 6639 assembly gloves are suitable for precision mechanical work in dry to slightly damp work environments. The robust polyurethane coating provides abrasion-resistance, without compromising dexterity or tactility.
  • Polyamide gloves with polyurethane coating on the palm and fingertips
  • Coating offers a high level of abrasion resistance
  • Enhanced grip capabilities in dry, or slightly damp applications
  • Lightweight and sensitive, ideal for precision mechanical work
  • Dark colored material hides dirt and stains
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Black
Exterior Finish Smooth
Hand Ambidextrous
Material Polyamide
Product Type Mechanical Protection Gloves
Size 6
Style Full-hand Mitt