Learn More
Uvex™ silv-Air Respirator FFP3
Product Code. 15355772 Shop All Uvex Products
Change view
Click to view available options
:
Silver/Anthracite with valve and carbon
Product Code.
15355772 Silver/Anthracite with valve and carbon
1 options
Product Code. 15355772 Supplier Uvex™ Supplier No. 8707320
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Particulate filtering form mask with exhalation valve - Protection level FFP3
  • Protection against pollutants up to 30 times the limit
  • Soft material edges for more comfort
  • Very soft, circumferential sealing lip for optimum comfort and reliable sealing seat
  • Integrated flexible adjustable nose clip
  • 4-point headband attachment for comfortable, individual and secure fit of the mask
  • Exhalation valve to reduce expiratory resistance and to reduce heat and moisture under the mask
  • Activated carbon filter against odors, gases and vapors below limit
  • Mask meets both the requirements for reusability (R) and the requirement for high dust exposure (D-marking, dolomite dust test)
  • Art. 8707.320
  • Packing unit: 5 pieces
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 149, meets both the requirements for reusability (R) and the requirement for high dust exposure (D-marking, dolomite dust test), FFP3 protection level
    Color Gray
    Filter Type Activated carbon filter
    Product Type Particulate filtering form mask with exhalation valve
    With Exhalation Valve Yes
    Quantity 5/Pk.
    Resistant To Pollutants up to 30 times the limit and odors, gases and vapors below limit