Learn More
Uvex™ silv-Air 2210 Respirator FFP2
Product Code. 15305792 Shop All Uvex Products
Change view
Click to view available options
:
White, with valve
Product Code.
15305792 White, with valve
1 options
Product Code. 15305792 Supplier Uvex™ Supplier No. 8732210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Particulate filtering form mask with exhalation valve - Protection level FFP2
  • Protection against pollutants up to 10 times the limit
  • Soft material edges for more comfort
  • Continuous headband for comfortable and secure fit of the mask
  • Exhalation valve to reduce the exhalation resistance and to reduce heat and moisture under the mask
  • Integrated, flexibly adaptable nose clip
  • Sealing lip in the nose area for high wearing comfort and reliable sealing seat
  • Compatibility with uvex goggles
  • Mask meets the requirements against high levels of dust (D-marking, dolomite dust test)
  • Art. 8732.210
  • Packing unit: 15 pieces
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 149, Meets the requirements against high levels of dust (D-marking, dolomite dust test, Protection level FFP2
    Color White
    Filter Type Classic cup style FFP2
    Product Type Particulate filtering form mask with exhalation valve
    With Exhalation Valve Yes
    Quantity 15/Pk.
    Resistant To Pollutants up to 10 times the limit
    With Faceseal Yes