missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ C500 Wet Plus Cut Protection Glove

Product Code. p-7160279 Shop All Uvex Products
Change view
Click to view available options
Size:
7
9
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15357052 7 Each
15377052 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15357052 Supplier Uvex™ Supplier No. 6049607
Add to basket
Add to basket

Suitable for wet areas and oily working conditions.

  • The use of Bamboo TwinFlex® Technology ensures a high level of cut resistance, an exceptional abrasion resistance and a supreme wearing comfort
  • The innovative SoftGrip coating guarantees an excellent dexterity, flexibility and secure grip
  • The uvex C500 wet plus model is certified in accordance with EN 407 for contact heat up to 100 °C
  • Very high cut protection (cut 5)
  • Silicon-free according to imprint test
  • Certified according to Öko-Tex Standard 100
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance EN 388:2016 (4 X 4 2 C), 407 (X 1 X X X X)
    Coated Area Back of the hand partially coated
    Coating Material HPE
    Color Lime Green, Anthracite
    Hand Ambidextrous
    Latex Free Yes
    Material HPPE, Polyamide
    Product Type Cut-resistant Gloves
    Size 7
    For Use With (Application) Automotive industry, glass industry, maintenance/servicing, metal work industry, assembly work, and transport industry
    Glove Cuff Style Knit
    Model C500 WET PLUS
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.