Learn More
uvex C300 Foam Cut Protection Gloves
Product Code. 15357092 Shop All Uvex Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
Product Code. 15357092
Brand: uvex 6054409
Only null left
Add to basket
Add to basket
Uvex C300 Foam gloves set standards for comfort, tactility and efficiency while also providing level 3/C cut protection. High-performance elastomer coating on the palm and fingertips enhances grip capabilities in dry or slightly damp applications.
  • Gloves provide level 3/C blade cut resistance, abrasion resistance, and high tear resistance
  • High-performance elastomer (HPE) coating on the palm and fingers for enhanced grip capabilities
  • Soft-grip foam coating provides mechanical abrasion resistance
  • Highly flexible with very good tactile sensation
  • Uvex Climazone technology reduces excessive moisture and warmth
  • 3D Ergo technology for a secure fit
  • Silicone-free according to the imprint test
TRUSTED_SUSTAINABILITY
Specifications
Coating Material HPE
Color Gray
Hand Ambidextrous
Latex Free Yes
Material Bamboo Viscose, HPPE, Polyamide, Glass
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 3X42C
Glove Cuff Style Knit
Series C300 Foam