missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The new generation G:BOX with high resolution camera and application controlled GeneSys software for all fluorescence requirements.
Gel documentation and analysis system
Gel documentation and analysis system
- Modular system
- Light tight darkroom - suitable for fluorescence, upgradeable for chemiluminescence
- 16bit 3.8MP camera with auto focus, auto exposure and series capture
- Integration control - capture faint bands or features from any gel
- Neutral fielding for background correction
- Filter selection options for detection of a wide range of stains and dyes
- GeneSys software includes a database of hundreds of applications
- The system includes a motor driven zoom lens as standard and a motor driven filter wheel
- Internal lighting comes from an optional UV transilluminator or a blue light LED model to further extend the applications possible with the G:BOX F3
- With unlimited copies of GeneTools analysis software
- Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver stain, SYBR™ Gold, SYBR™ Green, SYBR™ Safe, GelStar™, SYPRO™ Red , SYPRO™ Ruby, SYPRO™ Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond , Deep Purple™
- Extended fluorescence - for use with EPI RGB lighting module
- Agar plates
- Cells in flasks
- Autoradiographs
- Membranes
- Spots and slot blots
- TLC plates
- Microtitre plates
- Macro-arrays
Specifications
Specifications
| Intensity | Fixed |
| Wavelength | 254 nm, 365 nm |
| For Use With (Equipment) | G:BOX systems |
| Type | UV epi Light, Short And Long Wave |
Product Title
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
