missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568440
This item is not returnable.
View return policy
Description
UTY Polyclonal Antibody for Western Blot, IHC (P)
UTY encodes a protein containing tetratricopeptide repeats which are thought to be involved in protein-protein interactions. This protein is a minor histocompatibility antigen which may induce graft rejection of male stem cell grafts.
Specifications
| UTY | |
| Polyclonal | |
| Unconjugated | |
| UTY | |
| histone demethylase UTY; Hydb; Kdm6a; KDM6AL; KDM6C; lysine (K)-specific demethylase 6A; lysine-specific demethylase 6A; male-specific histocompatibility antigen H-YDB; mKIAA4057; RGD1565481; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat containing, Y-linked; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR gene on Y chromosome; ubiquitously transcribed TPR protein on the Y chromosome; ubiquitously transcribed Y chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; Utx; Uty; UTY1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22290, 7404 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O14607, P79457 | |
| UTY | |
| Full length Fusion protein of human UTY. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction