missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTY Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94768-0.02ml
This item is not returnable.
View return policy
Description
UTY Polyclonal antibody specifically detects UTY in Mouse, Rat samples. It is validated for Western Blot
Specifications
| UTY | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZp686L12190, EC 1.14.11, EC 1.14.11.-, histone demethylase UTY, tetratricopeptide repeat protein, ubiquitous TPR motif protein UTY, ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome, ubiquitously transcribed tetratricopeptide repeat gene, Y-linked, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 101, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 106, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 11, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 118, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 121, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 132, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 133, ubiquitously transcribed tetratricopeptide repeat protein Y-linked transcriptvariant 136, ubiquitou | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human UTY (NP_009056.3). MKSCAVSLTTAAVAFGDEAKKMAEGKASRESEEESVSLTVEEREALGGMDSRLFGFVRLHEDGARTKTLL | |
| 0.02 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7404 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction