Learn More
UTRN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27948-1-AP-150UL
This item is not returnable.
View return policy
Description
The UTRN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UTRN. This antibody recognizes human antigen. The UTRN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UTRN | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7402 | |
| IgG |
| Western Blot, ELISA | |
| 433 μg/mL | |
| Rabbit | |
| Primary | |
| Human |