Learn More
UTP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17671-1-AP-20UL
 View more versions of this product
Product Code. 16897604
  • £135.00 / 20µL
Description
This gene encodes glucose-6-phosphate dehydrogenase. This protein is a cytosolic enzyme encoded by a housekeeping X-linked gene whose main function is to produce NADPH, a key electron donor in the defense against oxidizing agents and in reductive biosynthetic reactions. G6PD is remarkable for its genetic diversity. Many variants of G6PD, mostly produced from missense mutations, have been described with wide ranging levels of enzyme activity and associated clinical symptoms. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
UTP6
Polyclonal
Unconjugated
UTP6
C17orf40, HCA66, MHAT, Multiple hat domains protein, UTP6
Rabbit
Antigen Affinity Chromatography
RUO
216987, 360574, 55813
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VCY6, Q9NYH9
Utp6
UTP6 Fusion Protein Ag11327
20 μL
Primary
Mouse, Human, Rat