Learn More
Invitrogen™ UTP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521716
Description
Recommended positive controls: HCT116. Predicted reactivity: Mouse (82%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. PA5-21716 has been used successfully in Northern Blot. Dilution is to be determined by the consumer.
Involved in nucleolar processing of pre-18S ribosomal RNA. [UniProt].
Specifications
| UTP6 | |
| Polyclonal | |
| Unconjugated | |
| UTP6 | |
| 4732497O03Rik; C17orf40; HCA66; hepatocellular carcinoma associated antigen 66; hepatocellular carcinoma-associated antigen 66; Mhat; Multiple hat domains protein; RGD1311017; U3 small nucleolar RNA-associated protein 6 homolog; Utp6; UTP6 small subunit processome component; UTP6, small subunit (SSU) processome component, homolog; UTP6, small subunit (SSU) processome component, homolog (yeast); UTP6, small subunit processome component | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NYH9 | |
| UTP6 | |
| Recombinant protein encompassing a sequence within the center region of human UTP6. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.