missing translation for 'onlineSavingsMsg'
Learn More

UTP6 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18407444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407444 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407444 Supplier Novus Biologicals Supplier No. NBP130467AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UTP6 Polyclonal antibody specifically detects UTP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C17orf40, chromosome 17 open reading frame 40, HCA66hepatocellular carcinoma associated antigen 66, Hepatocellular carcinoma-associated antigen 66, MHAT, Multiple hat domains protein, U3 small nucleolar RNA-associated protein 6 homolog, UTP6, small subunit (SSU) processome component, homolog (yeast)
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 565 and 597 of human hepatocellular carcinoma associated antigen 66 using the numbering given in entry NP_060898.2 (GeneID 55813).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55813
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.