missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP6 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30467AF405
This item is not returnable.
View return policy
Description
UTP6 Polyclonal antibody specifically detects UTP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| UTP6 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C17orf40, chromosome 17 open reading frame 40, HCA66hepatocellular carcinoma associated antigen 66, Hepatocellular carcinoma-associated antigen 66, MHAT, Multiple hat domains protein, U3 small nucleolar RNA-associated protein 6 homolog, UTP6, small subunit (SSU) processome component, homolog (yeast) | |
| The immunogen for this product maps to a region between residue 565 and 597 of human hepatocellular carcinoma associated antigen 66 using the numbering given in entry NP_060898.2 (GeneID 55813). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55813 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction