missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557354
This item is not returnable.
View return policy
Description
Immunogen sequence: LQEDDFGVAW VEAFAKPVPQ VDEAETRVVK DLAKVSVKEK LKMLRKESPE LLELIEDLKV KLTEVKDELE PLLELVEQGI IPPGKGSQ Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 95%.
Essential for gene silencing: has a role in the structure of silenced chromatin. Plays a role in the developing brain. [UniProt].
Specifications
| UTP3 | |
| Polyclonal | |
| Unconjugated | |
| Utp3 | |
| 2400011K09Rik; AW557551; C87704; charged amino acid rich leucine zipper 1; charged amino acid rich leucine zipper 1 homolog; charged amino acid-rich leucine zipper 1; Crl1; Crl-1; Crlz1; disrupter of silencing 10; disrupter of silencing SAS10; Sas10; something about silencing protein 10; UTP3; UTP3 homolog; UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae); UTP3, small subunit processome component; UTP3, small subunit processome component homolog (S. cerevisiae) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQZ2 | |
| Utp3 | |
| Recombinant protein corresponding to Human UTP3. Recombinant protein control fragment (Product #RP-96649). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction