missing translation for 'onlineSavingsMsg'
Learn More

UTP3 Antibody, Novus Biologicals™

Product Code. 18417128 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417128 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417128 Supplier Novus Biologicals Supplier No. H00057050B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

UTP3 Polyclonal antibody specifically detects UTP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Accession No. NP_065101.1
Gene Alias Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae)
Host Species Mouse
Immunogen UTP3 (NP_065101.1, 1 a.a. - 479 a.a.) full-length human protein. MVGRSRRRGAAKWAAVRAKAGPTLTDENGDDLGLPPSPGDTSYYQDQVDDFHEARSRAALAKGWNEVQSGDEEDGEEEEEEVLALDMDDEDDEDGGNAGEEEEEENADDDGGSSVQSEAEASVDPSLSWGQRKKLYYDTDYGSKSRGRQSQQEAEEEEREEEEEAQIIQRRLAQALQEDDFGVAWVEAFAKPVPQVDEAETRVVKDLAKVSVKEKLKMLRKESPELLELIEDLKVKLTEVKDELEPLLELVEQGIIPPGKGSQYLRTKYNLYLNYCSNISFYLILKARRVPAHGHPVIERLVTYRNLINKLSVVDQKLSSEIRHLLTLKDDAVKKELIPKAKSTKPKPKSVSKTSAAACAVTDLSDDSDFDEKAKLKYYKEIEDRQKLKRKKEENSTEEQALEDQNAKRAITYQIAKNRGLTPRRKKIDRNPRVKHREKFRRAKIRRRGQVREVRKEEQRYSGELSGIRAGVKKSIKLK
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 57050
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.