missing translation for 'onlineSavingsMsg'
Learn More

UTP3 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18425253 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425253 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425253 Supplier Novus Biologicals Supplier No. NBP119108AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UTP3 Polyclonal antibody specifically detects UTP3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTP3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Charged amino acid-rich leucine zipper 1, CRL1charged amino acid rich leucine zipper 1 homolog, CRLZ1UTP3 homolog, disrupter of silencing 10, Disrupter of silencing SAS10, FLJ23256, SAS10DKFZp761F222, something about silencing protein 10, UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae)
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 429 and 479 of human something about silencing protein 10 using the numbering given in entry NP_065101.1 (GeneID 57050).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 57050
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.