Learn More
Invitrogen™ UTP18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590468
Beschreibung
Immunogen sequence: RRLRQRNRLR LEEDKPAVER CLEELVFGDV ENDEDALLRR LRGPRVQEHE DSGDSEVENE AKGNFPPQKK PVWVDEEDED EEMVDMMNNR FRKDMMKNAS ESKLSKDNLK KRLKEEFQHA MGGVPAWAET TKRKTSSDDE SEEDEDDLLQ RTGNFISTST SLPRGILKMK NCQHANAERP TVARISSVQF H; Positive Samples: HeLa, Jurkat, Mouse brain, Mouse kidney, Mouse liver; Cellular Location: Nucleus, nucleolus.
UTP18 is involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
Spezifikation
| UTP18 | |
| Polyclonal | |
| Unconjugated | |
| UTP18 | |
| 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; U3 small nucleolar RNA-associated protein 18 homolog; Utp18; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit (SSU) processome component, homolog (yeast); UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50; Wdr50 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 217109, 51096 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5SSI6, Q9Y5J1 | |
| UTP18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-260 of human UTP18 (NP_0570852). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.