missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP18 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94790-0.02ml
This item is not returnable.
View return policy
Description
UTP18 Polyclonal antibody specifically detects UTP18 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UTP18 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CGI-48, U3 small nucleolar RNA-associated protein 18 homolog, UTP18, small subunit (SSU) processome component, homolog (yeast), WD repeat domain 50, WD repeat-containing protein 50, WDR50 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-260 of human UTP18 (NP_057085.2). RRLRQRNRLRLEEDKPAVERCLEELVFGDVENDEDALLRRLRGPRVQEHEDSGDSEVENEAKGNFPPQKKPVWVDEEDEDEEMVDMMNNRFRKDMMKNASESKLSKDNLKKRLKEEFQHAMGGVPAWAETTKRKTSSDDESEEDEDDLLQRTGNFISTSTSLPRGILKMKNCQHANAERPTVARISSVQFH | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51096 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction