missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UTP15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565386
This item is not returnable.
View return policy
Description
Immunogen sequence: LWDIPNSKEIL TFKEHSDYVR CGCASKLNPD LFITGSYDHT VKMFDARTSE SVLSVEHGQP VESVLLFPSG GLLVSAG Highest antigen sequence identity to the following orthologs - mouse 92%, rat 94%.
Ribosome biogenesis factor. Involved in nucleolar processing of pre-18S ribosomal RNA. Required for optimal pre-ribosomal RNA transcription by RNA polymerase I. [UniProt].
Specifications
| UTP15 | |
| Polyclonal | |
| Unconjugated | |
| UTP15 | |
| AW544865; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; NET21; RGD1310992; Src-associated protein SAW; U3 small nucleolar RNA-associated protein 15 homolog; U3 small nucleolar RNA-associated protein 15 homolog; LOW QUALITY PROTEIN: U3 small nucleolar RNA-associated protein 15 homolog; UniProtKB:Q8TED0}; UTP15; utp15 {ECO:0000250; UTP15 small subunit processome component; UTP15 SSU processome component; UTP15, small subunit processome component; UTP15, U3 small nucleolar ribonucleoprotein, homolog; UTP15, U3 small nucleolar ribonucleoprotein, homolog (S. cerevisiae); UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TED0 | |
| UTP15 | |
| Recombinant Human UTP15. Recombinant protein control fragment (Product #RP-104794). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction